1,2,3-Thiadiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

045861-5G 045861-1G 045861-500MG
101872-984EA 806.15 USD
101872-984 101872-982 101872-980
1,2,3-Thiadiazole-4-carboxylic acid ≥95%
1,2,3-Thiadiazole-4-carboxylic acid

Hazard code(s): Irritant

Formula: C₃H₂N₂O₂S
Melting Pt: 230…232 °C
MDL Number: MFCD00052099
CAS Number: 4100-13-4

Order Now

Specification Test Results

Melting point 230-232°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR