2-Methylisophthalonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-1,3-benzenedicarbonitrile2-Methyl-1,3-dicyanobenzene2-Methylbenzene-1,3-dicarbonitrile2,6-Dicyanotoluene

045555-1G 045555-5G
101871-448EA 350.77 USD
101871-448 101871-450
2-Methylisophthalonitrile ≥95%
2-Methylisophthalonitrile

Hazard code(s): Irritant

Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 136…138 °C
MDL Number: MFCD00012285
CAS Number: 2317-22-8

Order Now

Specification Test Results

Melting point 136-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR