N-(3-Nitrobenzoyl)morpholine ≥95%
Supplier: Matrix Scientific
Synonyms:
Morpholino(3-nitrophenyl)methanone
045484-500MG
045484-1G
045484-5G
101871-078EA
532.31
USD
101871-078
101871-074
101871-076
N-(3-Nitrobenzoyl)morpholine ≥95%
N-(3-Nitrobenzoyl)morpholine
Hazard code(s): Irritant
Formula:
C₁₁H₁₂N₂O₄ MW: 236.23 g/mol Melting Pt: 86…88 °C |
MDL Number:
MFCD00451515 CAS Number: 262162-90-3 |
Specification Test Results
Melting point | 86-88°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...