4-Bromothiophene-3-carbaldehyde ≥90%

Supplier: Matrix Scientific

045184-1G
101869-572EA 175.95 USD
101869-572
4-Bromothiophene-3-carbaldehyde ≥90%
4-Bromothiophene-3-carbaldehyde

Hazard code(s): Irritant

Formula: C₅H₃BrOS
MW: 191.05 g/mol
Boiling Pt: 96 °C (5mm)
MDL Number: MFCD01318960
CAS Number: 18791-78-1

Order Now

Specification Test Results

Boiling point 96°C/5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR