3-Isothiocyanatopyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Pyridyl isothiocyanate

044983-500MG 044983-5G 044983-1G
101868-570EA 806.15 USD
101868-570 101868-568 101868-566
3-Isothiocyanatopyridine ≥95%
3-Isothiocyanatopyridine

Hazard code(s): Irritant

Formula: C₆H₄N₂S
MW: 136.18 g/mol
Boiling Pt: 118 °C (20mm)
MDL Number: MFCD00052351
CAS Number: 17452-27-6

Order Now

Specification Test Results

Boiling point 118°C/20mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR