Methyl-4'-nitrobiphenyl-4-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4'-nitro[1,1'-biphenyl]-4-carboxylate
043966-500MG
043966-1G
043966-5G
101863-440EA
223.08
USD
101863-440
101863-444
101863-442
Methyl-4'-nitrobiphenyl-4-carboxylate ≥95%
Methyl 4'-nitrobiphenyl-4-carboxylate
Hazard code(s): Irritant
Formula:
C₁₄H₁₁NO₄ Melting Pt: 188…190 °C |
MDL Number:
MFCD04117797 CAS Number: 5730-75-6 |
Specification Test Results
Melting point | 188-190°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...