Methyl-6-aminonicotinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 6-aminonicotinate
, 6-Aminonicotinic acid methyl ester
043257-1G
043257-25G
043257-5G
101859-930EA
29.23
USD
101859-930
101859-932
101859-928
Methyl-6-aminonicotinate ≥95%
Methyl 6-aminonicotinate
Hazard code(s): Irritant
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol Melting Pt: 158…160 °C |
MDL Number:
MFCD00797844 CAS Number: 36052-24-1 |
Specification Test Results
Melting point | 158-160°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...