Methyl-5-bromo-2-chloronicotinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 5-bromo-2-chloronicotinate
042788-1G
042788-5G
101857-534EA
21.54
USD
101857-534
101857-532
Methyl-5-bromo-2-chloronicotinate ≥95%
Methyl 5-bromo-2-chloronicotinate
Hazard code(s): Irritant
Formula:
C₇H₅BrClNO₂ MW: 250.48 g/mol Melting Pt: 50…52 °C |
MDL Number:
MFCD03844848 CAS Number: 78686-79-0 |
Specification Test Results
Melting point | 50-52°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...