2-Fluoro-3-methoxypyridine ≥98%

Supplier: Matrix Scientific

041230-5G 041230-1G
101850-994EA 70.77 USD
101850-994 101850-996
2-Fluoro-3-methoxypyridine ≥98%
2-Fluoro-3-methoxypyridine

Hazard code(s): Irritant

Formula: C₆H₆FNO
MDL Number: MFCD09834126
CAS Number: 163234-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR