5-Fluoropicolinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-2-picolinic acid5-Fluoropyridine-2-carboxylic acid5-Fluoro-2-pyridinecarboxylic acid

041223-1G
101850-970EA 15.38 USD
101850-970
5-Fluoropicolinic acid ≥98%
5-Fluoropicolinic acid

Hazard code(s): Irritant

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
MDL Number: MFCD04114196
CAS Number: 107504-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR