(4-Chlorophenylthio)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-((4-Chlorophenyl)thio)acetic acid

040411-500MG
101849-458EA 37.95 USD
101849-458
(4-Chlorophenylthio)acetic acid
(4-Chlorophenylthio)acetic acid
Formula: C₈H₇ClO₂S
MW: 202.66 g/mol
MDL Number: MFCD00021762
CAS Number: 3405-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR