5-(4-(Diethylamino)benzylidene)rhodanine

Supplier: Matrix Scientific

Synonyms: (5E)-5-[4-(Diethylamino)benzylidene]-2-mercapto-1,3-thiazol-4(5H)-one

040156-500MG
101848-950EA 116.92 USD
101848-950
5-(4-(Diethylamino)benzylidene)rhodanine
5-(4-(Diethylamino)benzylidene)rhodanine
Formula: C₁₄H₁₆N₂OS₂
MDL Number: MFCD04079831
CAS Number: 35778-58-6
EINECS: 252-723-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR