2-(Benzyloxy)benzoic acid

Supplier: Matrix Scientific

Synonyms: 2-Benzyloxybenzoic acid

039992-500MG
101848-622EA 13.8 USD
101848-622
2-(Benzyloxy)benzoic acid
2-(Benzyloxy)benzoic acid
Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
MDL Number: MFCD00051940
CAS Number: 14389-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR