Ethyl indole-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Indole-2-Carboxylic Acid Ethyl Ester2-CarbethoxyinoleEthyl 1H-indole-2-carboxylate

038878-5G 038878-100G 038878-25G
101846-390EA 41.54 USD
101846-390 101846-392 101846-388
Ethyl indole-2-carboxylate ≥97%
Ethyl indole-2-carboxylate

Hazard code(s): Irritant

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
MDL Number: MFCD00005609
CAS Number: 3770-50-1
EINECS: 223-206-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR