2-Nitrophenyl isocyanate

Supplier: Matrix Scientific

Synonyms: 2-Nitrophenylisocyanate1-Isocyanato-2-nitrobenzene

038667-500MG
101845-972EA 193.85 USD
101845-972
2-Nitrophenyl isocyanate
2-Nitrophenyl isocyanate
Formula: C₇H₄N₂O₃
MW: 164.12 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007092
CAS Number: 3320-86-3
EINECS: 222-024-2
UN: 3335
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR