3-Amino-5-methylcaproic acid

Supplier: Matrix Scientific

Synonyms: 3-Amino-5-methylhexanoic acidDL-beta-Homoleucine(+/-)-3-Amino-5-methylhexanoic acid

038647-500MG
101845-930EA 30.77 USD
101845-930
3-Amino-5-methylcaproic acid
3-Amino-5-methylcaproic acid
Formula: C₇H₁₅NO₂
MW: 145.2 g/mol
Melting Pt: 227 °C
MDL Number: MFCD01863281
CAS Number: 3653-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR