2,5-Dimethoxycinnamic acid

Supplier: Matrix Scientific

Synonyms: 3-(2,5-Dimethoxyphenyl)acrylic acidtrans-2,5-Dimethoxycinnamic acid2',5'-Dimethoxycinnamic acid

038633-500MG
101845-900EA 146.15 USD
101845-900
2,5-Dimethoxycinnamic acid
2,5-Dimethoxycinnamic acid
Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 150 °C
MDL Number: MFCD00004378
CAS Number: 10538-51-9
EINECS: 234-114-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR