2,5-Dimethylphenyl isocyanate

Supplier: Matrix Scientific

Synonyms: 2-Isocyanato-1,4-dimethylbenzene2,5-Xylyl isocyanate

038307-500MG
101845-234EA 116.92 USD
101845-234
2,5-Dimethylphenyl isocyanate
2,5-Dimethylphenyl isocyanate
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 210 °C (1013 hPa)
Density: 1.043 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00013854
CAS Number: 40397-98-6
UN: 2206
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR