3-Chloro-4-methylphenyl isocyanate

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-isocyanato-1-methylbenzene

038305-500MG
101845-230EA 193.85 USD
101845-230
3-Chloro-4-methylphenyl isocyanate
3-Chloro-4-methylphenyl isocyanate
Formula: C₈H₆ClNO
MW: 167.59 g/mol
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 23.5 °C
Density: 1.224 g/cm³ (20 °C)
MDL Number: MFCD00013858
CAS Number: 28479-22-3
EINECS: 249-050-7
UN: 2236
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR