5-Benzyloxytryptamine hydrochloride

Supplier: Matrix Scientific

Synonyms: 2-[5-(Benzyloxy)-1H-indol-3-yl]ethanamine hydrochloride5-(phenylmethoxy)indole-3-ethanamine monohydrochloride

038028-500MG
101844-668EA 210.77 USD
101844-668
5-Benzyloxytryptamine hydrochloride
5-Benzyloxytryptamine hydrochloride

Hazard code(s): Irritant

Formula: C₁₇H₁₉ClN₂O
Melting Pt: 260…3 °C
MDL Number: MFCD00012685
CAS Number: 52055-23-9
EINECS: 257-626-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR