4-Hydroxyquinolin-2(1H)-one ≥95%

Supplier: Matrix Scientific
Warning

037714-100G
101844-022EA 267.69 USD
101844-022
4-Hydroxyquinolin-2(1H)-one ≥95%
4-Hydroxyquinolin-2(1H)-one

Hazard code(s): Irritant

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00277932
CAS Number: 86-95-3
EINECS: 201-711-0

Order Now

Specification Test Results

Melting point 300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR