2-Pyrimidinecarbonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Cyanopyrimidine

037606-100G
101843-780EA 72.31 USD
101843-780
2-Pyrimidinecarbonitrile ≥98%
2-Pyrimidinecarbonitrile

Hazard code(s): Irritant

Formula: C₅H₃N₃
MW: 105.1 g/mol
MDL Number: MFCD00160513
CAS Number: 14080-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR