4,5-Dibromo-2-thiophenesulfonyl chloride

Supplier: Matrix Scientific

Synonyms: 4,5-Dibromothiophene-2-sulphonyl chloride4,5-Dibromothiophene-2-sulfonyl chloride

037560-500MG
101843-662EA 193.85 USD
101843-662
4,5-Dibromo-2-thiophenesulfonyl chloride
4,5-Dibromo-2-thiophenesulphonyl chloride
Formula: C₄HBr₂ClO₂S₂
MW: 340.44 g/mol
Melting Pt: 80 °C
MDL Number: MFCD00041439
CAS Number: 81606-31-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR