1,2-Benzenedisulfonyl chloride
Supplier: Matrix Scientific
Synonyms:
Benzene-1,2-di(sulphonyl chloride)
, Benzene-1,2-di(sulfonyl chloride)
037508-500MG
101843-560EA
146.15
USD
101843-560
1,2-Benzenedisulfonyl chloride
1,2-Benzenedisulphonyl chloride
Formula:
C₆H₄Cl₂O₄S₂ MW: 275.13 g/mol |
MDL Number:
MFCD00059836 CAS Number: 6461-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...