trans-3-Chlorocinnamic acid

Supplier: Matrix Scientific
Warning

Synonyms: (E)-3-(3-Chlorophenyl)acrylic acid

037479-500MG
101843-492EA 116.92 USD
101843-492
trans-3-Chlorocinnamic acid
trans-3-Chlorocinnamic acid
Formula: C₉H₇ClO₂
MW: 182.6037 g/mol
MDL Number: MFCD00004384
CAS Number: 14473-90-6
EINECS: 217-478-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR