2-Aminobenzylamine

Supplier: Matrix Scientific

Synonyms: 2-(Aminomethyl)aniline

037471-500MG
101843-466EA 15.38 USD
101843-466
2-Aminobenzylamine
2-Aminobenzylamine
Formula: C₇H₁₀N₂
MDL Number: MFCD00075408
CAS Number: 4403-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR