3-Furoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Furancarboxylic acidFuran-3-carboxylic acid

037461-100G
101843-446EA 558.46 USD
101843-446
3-Furoic acid ≥97%
3-Furoic acid

Hazard code(s): Irritant

Formula: C₅H₄O₃
MW: 112.08 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD00005350
CAS Number: 488-93-7
EINECS: 207-689-9

Order Now

Specification Test Results

Melting point 122-124

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR