1-Hexylpiperazine

Supplier: Matrix Scientific

037425-500MG
101843-346EA 193.85 USD
101843-346
1-Hexylpiperazine
1-Hexylpiperazine
Formula: C₁₀H₂₂N₂
MW: 170.3 g/mol
MDL Number: MFCD01075185
CAS Number: 51619-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR