3-Bromo-5-fluorobenzaldehyde

Supplier: Matrix Scientific

037416-5G
101843-328EA 58.46 USD
101843-328
3-Bromo-5-fluorobenzaldehyde
3-Bromo-5-fluorobenzaldehyde
Formula: C₇H₄BrFO
MDL Number: MFCD04116319
CAS Number: 188813-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR