2,4,6-Trimethylbenzylamine

Supplier: Matrix Scientific

Synonyms: 1-MesitylmethanamineMesitylmethanamine

037106-500MG
101842-690EA 93.85 USD
101842-690
2,4,6-Trimethylbenzylamine
2,4,6-Trimethylbenzylamine
Formula: C₁₀H₁₅N
MW: 149.24 g/mol
MDL Number: MFCD03411013
CAS Number: 40393-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR