N,N-Bis(2-hydroxyethyl)-3-chloraniline

Supplier: Matrix Scientific

Synonyms: 2-[(3-Chlorophenyl)(2-hydroxyethyl)amino]ethanol

037068-500MG
101842-606EA 193.85 USD
101842-606
N,N-Bis(2-hydroxyethyl)-3-chloraniline
N,N-Bis(2-hydroxyethyl)-3-chloraniline
Formula: C₁₀H₁₄ClNO₂
MDL Number: MFCD00002852
CAS Number: 92-00-2
EINECS: 202-115-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR