5-Fluorotryptamine hydrochloride

Supplier: Matrix Scientific

Synonyms: 2-(5-Fluoro-1H-indol-3-yl)ethanamine hydrochloride

036773-500MG
101842-052EA 109.23 USD
101842-052
5-Fluorotryptamine hydrochloride
5-Fluorotryptamine hydrochloride
Formula: C₁₀H₁₂ClFN₂
MDL Number: MFCD00051081
CAS Number: 2711-58-2
EINECS: 220-307-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR