3',4'-Dimethylacetophenone

Supplier: Matrix Scientific

Synonyms: 1-(3,4-Dimethylphenyl)ethan-1-one1-(3,4-Dimethylphenyl)ethanone

036403-500MG
101841-290EA 387.69 USD
101841-290
3',4'-Dimethylacetophenone
3',4'-Dimethylacetophenone
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: –3 °C
Density: 1.0185 g/cm³ (0 °C)
MDL Number: MFCD00008743
CAS Number: 3637-01-2
EINECS: 222-859-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR