3-Hydrazinobenzoic acid
Supplier: Matrix Scientific
035978-100G
035978-25G
101840-448EA
103.08
USD
101840-448
101840-450
3-Hydrazinobenzoic acid
3-Hydrazinobenzoic acid
Hazard code(s): Irritant, Refrigerate
Formula:
C₇H₈N₂O₂ Melting Pt: ∼194 °C (dec.) |
MDL Number:
MFCD00191447 CAS Number: 38235-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...