2,5-Dichloroisonicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2,5-Dichloropyridine-4-carboxylic acid

035659-25G 035659-5G
101839-592EA 47.69 USD
101839-592 101839-594
2,5-Dichloroisonicotinic acid ≥98%
2,5-Dichloroisonicotinic acid

Hazard code(s): Irritant

Formula: C₆H₃Cl₂NO₂
MDL Number: MFCD00466640
CAS Number: 88912-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR