4-Chloro-3-fluorophenylboronic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
(4-Chloro-3-fluorophenyl)boronic acid
, Dihydroxy(4-chloro-3-fluorophenyl)borane
, 4-Chloro-3-fluorobenzeneboronic acid
, (4-Chloro-3-fluorophenyl)metaboric acid
035575-5G
035575-25G
101839-532EA
92.31
USD
101839-532
101839-534
4-Chloro-3-fluorophenylboronic acid ≥95%
4-Chloro-3-fluorophenylboronic acid
Hazard code(s): Irritant, Refrigerate
Formula:
C₆H₅BClFO₂ MW: 174.37 g/mol |
MDL Number:
MFCD01319010 CAS Number: 137504-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...