2-Formyl-4-thiopheneboronic acid

Supplier: Matrix Scientific

Synonyms: 2-Formylthiophene-4-boronic acid

035464-1G 035464-5G
101839-438EA 44.62 USD
101839-438 101839-440
2-Formyl-4-thiopheneboronic acid
2-Formyl-4-thiopheneboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₅H₅BO₃S
MW: 155.97 g/mol
Melting Pt: 270…272 °C
MDL Number: MFCD03002365
CAS Number: 175592-59-3

Order Now

Specification Test Results

Melting point 270-272

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR