2-Chloro-3-pyridineboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloropyridine-3-boronic acid2-Chloro-3-pyridinylboronic acid(2-Chloropyridin-3-yl)boronic acid

035405-5G
101839-406EA 53.85 USD
101839-406
2-Chloro-3-pyridineboronic acid ≥95%
2-Chloro-3-pyridineboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
MDL Number: MFCD03094997
CAS Number: 381248-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR