2,3-Dimethoxy-5-pyridineboronic acid pinacol ester

Supplier: Matrix Scientific

Synonyms: 2,3-Dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

035212-250MG
101839-202EA 455.4 USD
101839-202
2,3-Dimethoxy-5-pyridineboronic acid pinacol ester
2,3-Dimethoxy-5-pyridineboronic acid pinacol ester
Formula: C₁₃H₂₀BNO₄
MDL Number: MFCD11857651
CAS Number: 1083168-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR