4-Methylbenzamidoxime ≥97%
Supplier: Matrix Scientific
Synonyms:
N-Hydroxy-4-methylbenzimidamide
034855-5G
034855-25G
101838-516EA
936.92
USD
101838-516
101838-514
4-Methylbenzamidoxime ≥97%
4-Methylbenzamidoxime
Hazard code(s): Irritant
Formula:
C₈H₁₀N₂O MW: 150.18 g/mol Melting Pt: 142…146 °C |
MDL Number:
MFCD00019952 CAS Number: 19227-13-5 |
Specification Test Results
Melting point | 142-146°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...