5-Chloro-2-pyridineboronic acid ≥98%

034742-1G
101838-262EA 298.35 USD
101838-262
5-Chloro-2-pyridineboronic acid ≥98%
5-Chloro-2-pyridineboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
MDL Number: MFCD07368862
CAS Number: 652148-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR