1-(2-Bromoethoxy)-3-(trifluoromethyl)benzene

Supplier: Matrix Scientific

Synonyms: 3-(2-Bromoethoxy)benzotrifluoride

033164-500MG
101835-132EA 55.38 USD
101835-132
1-(2-Bromoethoxy)-3-(trifluoromethyl)benzene
1-(2-Bromoethoxy)-3-(trifluoromethyl)benzene
Formula: C₉H₈BrF₃O
MW: 269.06 g/mol
MDL Number: MFCD00215501
CAS Number: 18800-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR