4-Methylpyrimidine ≥97%

Supplier: Matrix Scientific

032435-25G
101833-722EA 67.69 USD
101833-722
4-Methylpyrimidine ≥97%
4-Methylpyrimidine

Hazard code(s): Flammable

Formula: C₅H₆N₂
Boiling Pt: 141…142 °C
Melting Pt: 32 °C
Density: 1.030 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00006115
CAS Number: 3438-46-8
EINECS: 222-344-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 141-142°C
Density 1.03
ND20 1.496

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR