4-Amino-3-iodopyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Iodopyridin-4-amine3-Iodo-4-pyridylamine

031253-5G 031253-1G
101831-316EA 21.54 USD
101831-316 101831-318
4-Amino-3-iodopyridine ≥98%
4-Amino-3-iodopyridine

Hazard code(s): Irritant

Formula: C₅H₅IN₂
MW: 220.01 g/mol
MDL Number: MFCD02663853
CAS Number: 88511-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR