4-(Diethylamino)butanone

Supplier: Matrix Scientific
Danger

Synonyms: 4-(Diethylamino)butan-2-one

030738-500MG
101830-734EA 193.85 USD
101830-734
4-(Diethylamino)butanone
4-(Diethylamino)butanone
Formula: C₈H₁₇NO
MW: 143.23 g/mol
Boiling Pt: 189 °C (1013 hPa)
Density: 0.86 g/cm³ (25 °C)
MDL Number: MFCD00009056
CAS Number: 3299-38-5
EINECS: 221-970-3
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR