1-Pentylpiperazine

Supplier: Matrix Scientific

029387-500MG
101828-050EA 217.35 USD
101828-050
1-Pentylpiperazine
1-Pentylpiperazine
Formula: C₉H₂₀N₂
MW: 156.27 g/mol
MDL Number: MFCD02093555
CAS Number: 50866-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR