Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(3,4-Dichlorobenzyl)piperazine

Supplier: Matrix Scientific


028765-500MG
101826-802EA 34 USD InStock
101826-802
1-(3,4-Dichlorobenzyl)piperazine
1-(3,4-Dichlorobenzyl)piperazine


Formula: C₁₁H₁₄Cl₂N₂
MW: 245.15 g/mol
MDL Number: MFCD03407488
CAS Number: 55513-17-2
EINECS: 000-000-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty