(±)-3,3-Dimethyl-2-butylamine

Supplier: Matrix Scientific

Synonyms: 1,2,2-Trimethylpropylamine

028756-500MG
101826-782EA 193.85 USD
101826-782
(±)-3,3-Dimethyl-2-butylamine
(±)-3,3-Dimethyl-2-butylamine
Formula: C₆H₁₅N
MW: 101.19 g/mol
MDL Number: MFCD00008078
CAS Number: 3850-30-4
EINECS: 223-353-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR