4-Acetoxy-3-methoxybenzaldehyde

Supplier: Matrix Scientific

Synonyms: Vanillin acetate4-Formyl-2-methoxyphenyl acetate

028689-100G
101826-646EA 235.38 USD
101826-646
4-Acetoxy-3-methoxybenzaldehyde
4-Acetoxy-3-methoxybenzaldehyde

Hazard code(s): Irritant

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Melting Pt: 76…78 °C
MDL Number: MFCD00003362
CAS Number: 881-68-5
EINECS: 212-920-1

Order Now

Specification Test Results

Melting point 76-78

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR