1-(4-Bromobenzyl)piperazine

Supplier: Matrix Scientific

027772-500MG
101824-826EA 193.85 USD
101824-826
1-(4-Bromobenzyl)piperazine
1-(4-Bromobenzyl)piperazine
Formula: C₁₁H₁₅BrN₂
MDL Number: MFCD02177416
CAS Number: 91345-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR